missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(3,5-dimethyl-1-benzothiophen-2-yl)-1-ethanone, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 388088-83-3 |
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Molecular Formula | C12H11BrOS |
MDL Number | MFCD03407327 |
Quantity | 1 g |
Synonym | 2-bromo-1-3,5-dimethyl-1-benzothiophen-2-yl-1-ethanone, 2-bromo-1-3,5-dimethyl-1-benzothiophen-2-yl ethanone, 2-bromo-1-3,5-dimethylbenzo b thiophen-2-yl ethanone, ethanone,2-bromo-1-3,5-dimethylbenzo b thien-2-yl, 2-bromo-1-3,5-dimethyl-1-benzothiophen-2-yl ethan-1-one |
InChI Key | BEGKHWMWXSYGJZ-UHFFFAOYSA-N |
SMILES | CC1=CC2=C(C=C1)SC(=C2C)C(=O)CBr |
IUPAC Name | 2-bromo-1-(3,5-dimethyl-1-benzothiophen-2-yl)ethanone |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'