missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-hydroxypyridine, 99%

Product Code. 11460583
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460583 5 g 5g
11470583 25 g 25g
11480583 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460583 Supplier Thermo Scientific Alfa Aesar Supplier No. A12687.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6602-32-0 | C5H4BrNO | 174.00 g/mol

2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6602-32-0
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00006220
InChI Key YKHQFTANTNMYPP-UHFFFAOYSA-N
Synonym 2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
PubChem CID 23070
IUPAC Name 2-bromopyridin-3-ol
SMILES OC1=CC=CN=C1Br

Specifications

Melting Point 182°C to 186°C
Quantity 5 g
Beilstein 109829
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 174
Percent Purity 99%
Chemical Name or Material 2-Bromo-3-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.