missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxybenzyl Alcohol, TRC

Product Code. 30134234
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134234 10 g 10g
17991675 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134234 Supplier Toronto Research Chemicals Supplier No. TRCB68463510G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54370-00-2
Molecular Formula C9 H11 Br O3
Molecular Weight (g/mol) 247.09
Synonym (2-Bromo-4,5-dimethoxyphenyl)methanol, 6-Bromoveratryl Alcohol, 2-Bromo-4,5-dimethoxybenzenemethanol;
IUPAC Name (2-bromo-4,5-dimethoxyphenyl)methanol
SMILES COc1cc(Br)c(CO)cc1OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H11BrO3/c1-12-8-3-6(5-11)7(10)4-9(8)13-2/h3-4,11H,5H2,1-2H3
Formula Weight 245.9892
Chemical Name or Material 2-Bromo-4,5-dimethoxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.