missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific Chemicals

Product Code. 15454257
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454257 5 g 5g
15444257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454257 Supplier Thermo Scientific Alfa Aesar Supplier No. H31649.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 351003-63-9 | Molecular Formula: C7H5BrF3NO2S | Molecular Weight: 304.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-63-9
Molecular Formula C7H5BrF3NO2S
Molecular Weight (g/mol) 304.081
MDL Number MFCD03094287
InChI Key IJUIBMWZUXWVLI-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl benzenesulfonamide, 2-bromo-4-trifluoromethyl benzenesulphonamide, 2-bromo-4-trifluoromethyl benzene sulfonamide, benzenesulfonamide,2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl benzene-1-sulfonamide, acmc-20ao2t, 2-bromanyl-4-trifluoromethyl benzenesulfonamide
PubChem CID 2778398
IUPAC Name 2-bromo-4-(trifluoromethyl)benzenesulfonamide
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)S(=O)(=O)N

Specifications

Melting Point 160°C to 164°C
Quantity 5 g
Formula Weight 304.1
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)benzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.