missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-5-fluorophenyl isothiocyanate, Thermo Scientific™

Product Code. 10260983
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10260983 10 g 10g
10205402 1 g 1g
10312315 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10260983 Supplier Thermo Scientific Maybridge Supplier No. SB01713EA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-35-3
Molecular Formula C7H3BrFNS
Molecular Weight (g/mol) 232.07
MDL Number MFCD00673194
InChI Key SSLVXFSUADEZBQ-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorophenyl isothiocyanate, 2-bromo-4-fluorophenylisothiocyanate, pubchem5018, acmc-20ao2g, 2-bromo-4-fluorobenzenisothiocyanate, benzene,1-bromo-4-fluoro-2-isothiocyanato, 2-bromo-4-fluorophenyl isothiocyanate, tech., benzene, 2-bromo-4-fluoro-1-isothiocyanato, 2-bromanyl-4-fluoranyl-1-isothiocyanato-benzene, benzene, 2-bromo-4-fluoro-1-isothiocyanato-9ci
PubChem CID 2736332
IUPAC Name 2-bromo-4-fluoro-1-isothiocyanatobenzene
SMILES FC1=CC=C(N=C=S)C(Br)=C1

Specifications

Quantity 10 g
Formula Weight 232.08
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-5-fluorophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.