Learn More
2-Bromo-5-nitroaniline, 98%, Thermo Scientific™
Product Code. 11387755
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387755 5 g 5g
11377755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11387755 Supplier Thermo Scientific Alfa Aesar Supplier No. L02597.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10403-47-1
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD00051578
InChI Key BAAUCXCLMDAZEL-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-nitro, 2-bromo-5-nitrobenzenamine, 3-amino-4-bromonitrobenzene, 2-bromo-5-nitro-phenylamine, 2-bromo-5-nitrophenylamine, pubchem21903, 2-bromo 5-nitroaniline, 2-bromo-5-nitro-aniline, 2-bromo-5-nitro aniline, acmc-2098ai
PubChem CID 82607
IUPAC Name 2-bromo-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)Br
Specifications
Melting Point 137°C to 141°C
Quantity 5 g
UN Number UN2811
Beilstein 2803492
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitroaniline