missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)phenol, 98%

Product Code. 15411029
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411029 25 g 25g
15401029 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15411029 Supplier Thermo Scientific Alfa Aesar Supplier No. H64009.14
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 402-05-1 | C7H4BrF3O | 241.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-05-1
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD09261190
InChI Key INHVNZLKNPJCJD-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl phenol, phenol, 2-bromo-5-trifluoromethyl, acmc-209jbu, phenol,2-bromo-5-trifluoromethyl, 3-bromo-5-hydroxybenzotrifluoride, 4-bromo-3-hydroxybenzotrifluoride
PubChem CID 21914470
IUPAC Name 2-bromo-5-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)O)Br

Specifications

Color Brown
Quantity 25 g
UN Number UN3265
Formula Weight 241.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.