missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-p-xylene, 98+%

Product Code. 11466927
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466927 25 g 25g
11476927 100 g 100g
11486927 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466927 Supplier Thermo Scientific Alfa Aesar Supplier No. A14358.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 553-94-6 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-94-6
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000074
InChI Key QXISTPDUYKNPLU-UHFFFAOYSA-N
Synonym 2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
PubChem CID 11121
IUPAC Name 2-bromo-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(Br)=C1

Specifications

Melting Point 9°C to 10°C
Density 1.347
Quantity 25 g
Refractive Index 1.551
Beilstein 2040350
Boiling Point 199°C to 201°C
Formula Weight 185.07
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-p-xylene
compliance-icons
Product Identifier
  • 2-Bromo-p-xylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-p-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.