missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo (S)-Carbidopa, TRC

Product Code. 30010467
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010467 25 mg 25mg
30005974 1 mg 1mg
30008998 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010467 Supplier Toronto Research Chemicals Supplier No. TRCB68206525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43197-33-7
Molecular Formula C10H13BrN2O4
Molecular Weight (g/mol) 305.13
Synonym (S)-2-Bromo-alpha-hydrazino-4,5-dihydroxy-alpha-methylbenzenepropanoic Acid
IUPAC Name (2S)-3-(2-bromo-4,5-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid
SMILES C[C@@](Cc1cc(O)c(O)cc1Br)(NN)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13BrN2O4/c1-10(13-12,9(16)17)4-5-2-7(14)8(15)3-6(5)11/h2-3,13-15H,4,12H2,1H3,(H,16,17)/t10-/m0/s1
Formula Weight 304.0059
Chemical Name or Material 2-Bromo (S)-Carbidopa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.