missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-5-(trifluoromethyl)furan, TRC

Product Code. 30338392
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30338392

Brand: Toronto Research Chemicals TRCB6879251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Bromomethyl)-5-(trifluoromethyl)furan
CAS 17515-77-4
Molecular Formula C6H4BrF3O
Quantity 1 g
Synonym 2-(Bromomethyl)-5-(trifluoromethyl)furan (ACI)
InChI Formula InChI=1S/C6H4BrF3O/c7-3-4-1-2-5(11-4)6(8,9)10/h1-2H,3H2
SMILES BrCC1=CC=C(O1)C(F)(F)F
IUPAC Name 2-(bromomethyl)-5-(trifluoromethyl)furan
Molecular Weight (g/mol) 228.99
Formula Weight 227.94
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.