missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl isothiocyanate, 98%

Product Code. 11352377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352377 1 g 1g
11362377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352377 Supplier Thermo Scientific Alfa Aesar Supplier No. L09971.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13037-60-0 | C7H4BrNS | 214.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13037-60-0
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004799
InChI Key PAFORXDSYWMYGP-UHFFFAOYSA-N
PubChem CID 611654
IUPAC Name 1-bromo-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Br

Specifications

Density 1.591
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6843
Quantity 1 g
UN Number UN2922
Beilstein 2802536
Sensitivity Moisture sensitive
Synonym 2-bromophenyl isothiocyanate, benzene,1-bromo-2-isothiocyanato, benzene, 1-bromo-2-isothiocyanato, 2-bromophenylisothiocyanate, benzene, bromoisothiocyanato, isothiocyanic acid 2-bromophenyl ester, acmc-1bbbv, o-bromophenyl isothiocyanate, 1-bromo-2-isothiocyanato-benzene, #
Formula Weight 214.09
Percent Purity 98%
Chemical Name or Material 2-Bromophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.