missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30317631
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317631 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317631 Supplier Dr. Ehrenstorfer Supplier No. DREA10862000AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butanone
CAS 78-93-3
Analyte or Component Names 2-Butanone
Shipping Condition Room Temperature
Molecular Formula C4 H8 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Butanon, Ethylmethylketon, Methylethylketon, MEK
InChI Formula InChI=1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3
SMILES CCC(=O)C
IUPAC Name butan-2-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 72.11
Quantity 1 mL
Formula Weight 72.0575 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.