missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethyl Acetate, TRC

Product Code. 30337013
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337013 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337013 Supplier Toronto Research Chemicals Supplier No. TRCB28172010ML
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butoxyethylacetate
CAS 112-07-2
Molecular Formula C8 H16 O3
Quantity 10 mL
Synonym Ethanol, 2-butoxy-, acetate (6CI,7CI,8CI,9CI), 2-Butoxyethyl acetate, B 0700, BCA, BMGAC, Butoxyethyl acetate, Butyl cellosolve acetate, Butyl glycol acetate, Dowanol EB acetate, EB Acetate, Ethylene glycol acetate monobutyl ether, Ethylene glycol butyl ether acetate, Ethylene glycol monobutyl ether acetate, Glycol monobutyl ether acetate
InChI Formula InChI=1S/C8H16O3/c1-3-4-5-10-6-7-11-8(2)9/h3-7H2,1-2H3
SMILES CCCCOCCOC(=O)C
IUPAC Name 2-butoxyethyl acetate
Molecular Weight (g/mol) 160.21
Formula Weight 160.1099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.