Learn More
2-Butoxyquinoline-4-carboxylic Acid, TRC
Product Code. 30333277
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333277 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30333277 Supplier Toronto Research Chemicals Supplier No. TRCB690340500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Butoxyquinoline-4-carboxylicAcid
CAS 10222-61-4
Molecular Formula C14 H15 N O3
Quantity 500 mg
Synonym 4-Quinolinecarboxylic acid, 2-butoxy-, Cinchoninic acid, 2-butoxy- (7CI,8CI), 2-Butoxy-4-quinolinecarboxylic Acid
InChI Formula InChI=1S/C14H15NO3/c1-2-3-8-18-13-9-11(14(16)17)10-6-4-5-7-12(10)15-13/h4-7,9H,2-3,8H2,1H3,(H,16,17)
SMILES CCCCOc1cc(C(=O)O)c2ccccc2n1
IUPAC Name 2-butoxyquinoline-4-carboxylic acid
Molecular Weight (g/mol) 245.27
Formula Weight 245.1052