missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxyphenylboronic Acid Pinacol Ester, TRC

Product Code. 30368543
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30368543 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368543 Supplier Toronto Research Chemicals Supplier No. TRCC181388250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1187591-17-8
Molecular Formula C13H17BO4
Quantity 250 mg
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic Acid
InChI Formula InChI=1S/C13H17BO4/c1-12(2)13(3,4)18-14(17-12)10-8-6-5-7-9(10)11(15)16/h5-8H,1-4H3,(H,15,16)
SMILES O=C(O)C=1C=CC=CC1B2OC(C)(C)C(O2)(C)C
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Weight (g/mol) 248.08
Formula Weight 248.12
Chemical Name or Material 2-Carboxyphenylboronic Acid Pinacol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.