missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-difluorobenzene, 98%, Thermo Scientific™

Product Code. 11492237
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492237 25 g 25g
11482237 5 g 5g
11454180 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492237 Supplier Thermo Scientific Alfa Aesar Supplier No. A14178.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2367-91-1
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042571
InChI Key XWCKSJOUZQHFKI-UHFFFAOYSA-N
PubChem CID 137561
IUPAC Name 2-chloro-1,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)F

Specifications

Density 1.352
Boiling Point 126°C to 128°C
Flash Point 20°C (68°F)
Odor Aromatic
Refractive Index 1.479
Quantity 25 g
UN Number UN1993
Beilstein 2081082
Synonym 1-chloro-2,5-difluorobenzene, 2,5-difluorochlorobenzene, benzene,2-chloro-1,4-difluoro, benzene, 2-chloro-1,4-difluoro, pubchem8529, acmc-20ai5u, ksc493a9f, 2-chloro-1,4-difluoro-benzene, 2-chloro-4-fluoro-fluorobenzene, 2-chloranyl-1,4-bis fluoranyl benzene
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-1,4-difluorobenzene
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.