Learn More
2-Chloro-2-methylnonane, 90+%
Product Code. 11310428
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310428 25 g 25g
11300428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11310428 Supplier Thermo Scientific Alfa Aesar Supplier No. 043784.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
CAS: 4325-50-2 | C10H21Cl | 176.728 g/mol
It is used in the photoelectron spectroscopy of C60Fn- and C60Fm2 in the gas phase and the generation and characterization of C1-C60F47- and D2-C60F44 in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4325-50-2
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.728
MDL Number MFCD00060781
InChI Key QDJXQCKHBZEIJM-UHFFFAOYSA-N
Synonym dimethyloctyl chloride, acmc-20ao0l, 2-chloro-2-methyl-nonane, nonane,2-chloro-2-methyl
PubChem CID 138189
IUPAC Name 2-chloro-2-methylnonane
SMILES CCCCCCCC(C)(C)Cl
Specifications
Color Colorless
Quantity 25 g
UN Number UN3082
Formula Weight 176.73
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylnonane