missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine, 97%

Product Code. 11364416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364416 1 g 1g
11374416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364416 Supplier Thermo Scientific Alfa Aesar Supplier No. L20022.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17282-04-1 | C5H3ClFN | 131.534 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 17282-04-1
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD03095302
InChI Key SVAZIMBLBHOVIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
PubChem CID 2782791
IUPAC Name 2-chloro-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Cl)F

Tekniske data

Density 1.325
Boiling Point 62°C to 64°C (23 mmHg)
Flash Point 72°C (161°F)
Odor Unpleasant
Refractive Index 1.5095
Quantity 1 g
UN Number UN2810
Beilstein 1363435
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-fluoropyridine
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.