missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine, 97%

Product Code. 11374416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374416 5 g 5g
11364416 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374416 Supplier Thermo Scientific Alfa Aesar Supplier No. L20022.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17282-04-1 | C5H3ClFN | 131.534 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17282-04-1
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD03095302
InChI Key SVAZIMBLBHOVIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
PubChem CID 2782791
IUPAC Name 2-chloro-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Cl)F

Specifications

Density 1.325
Boiling Point 62°C to 64°C (23 mmHg)
Flash Point 72°C (161°F)
Odor Unpleasant
Refractive Index 1.5095
Quantity 5 g
UN Number UN2810
Beilstein 1363435
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-fluoropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.