missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylquinoline, 97%, ACROS Organics™

Product Code. 10753251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753251 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10753251 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 268890050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 57876-69-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 84.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C10H8ClN
MDL Number MFCD00100265
Quantity 5 g
Synonym 2-chloro-3-methyl-quinoline, quinoline, 2-chloro-3-methyl, zlchem 105, pubchem12431, acmc-1ayrn, methyl 2-chloroquinoline, 2-chloro-3-methylquinolin, 3-methyl-2-chloroquinoline, 2-cloro-3-methyl-quinoline, 2-chloro-3-methyl quinoline
InChI Key UMYPPRQNLXTIEQ-UHFFFAOYSA-N
SMILES CC1=CC2=CC=CC=C2N=C1Cl
IUPAC Name 2-chloro-3-methylquinoline
Molecular Weight (g/mol) 177.631
PubChem CID 339082
Formula Weight 177.63
Percent Purity 97%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material 2-Chloro-3-methylquinoline, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.