missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dinitroaniline, 97%, Thermo Scientific Chemicals

Product Code. 11415817
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415817 100 g 100g
11425817 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415817 Supplier Thermo Scientific Alfa Aesar Supplier No. B24379.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3531-19-9
Molecular Formula C6H4ClN3O4
Molecular Weight (g/mol) 217.57
MDL Number MFCD00007147
InChI Key LHRIICYSGQGXSX-UHFFFAOYSA-N
Synonym 6-chloro-2,4-dinitroaniline, 2-chloro-4,6-dinitrophenyl amine, benzenamine, 2-chloro-4,6-dinitro, ccris 3112, aniline, 6-chloro-2,4-dinitro, 6-chloro-2,4-dinitrophenylamine, acmc-209ig0, 2,4-dinitro-6-chloroaniline, 3-12-00-01718 beilstein handbook reference, aronis23887
PubChem CID 19052
IUPAC Name 2-chloro-4,6-dinitroaniline
SMILES NC1=C(Cl)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 153°C to 155°C
Quantity 100 g
UN Number UN2811
Formula Weight 217.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4,6-dinitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.