missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoroanisole, 97%, Thermo Scientific™

Product Code. 10693415 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10693415 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10693415 Supplier Thermo Scientific Acros Supplier No. 426290050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2267-25-6
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 197.0°C to 200.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H6ClFO
Linear Formula ClC6H3(F)OCH3
MDL Number MFCD00672965
Quantity 5 g
Specific Gravity 1.29
Synonym 2-chloro-4-fluoroanisole, benzene, 2-chloro-4-fluoro-1-methoxy, 2-chloro-4-fluoro-1-methoxy-benzene, pubchem2658, acmc-1cnak, intermediates-zcf02609, ksc497i7l
InChI Key RKCGJVGMRPKPNY-UHFFFAOYSA-N
SMILES COC1=CC=C(F)C=C1Cl
IUPAC Name 2-chloro-4-fluoro-1-methoxybenzene
Molecular Weight (g/mol) 160.57
PubChem CID 2773580
Formula Weight 160.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-4-fluoroanisole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-fluoroanisole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.