missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoic acid, 98%

Product Code. 11447147
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447147 25 g 25g
11437147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447147 Supplier Thermo Scientific Alfa Aesar Supplier No. A14953.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2252-51-9 | C7H4ClFO2 | 174.555 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2252-51-9
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00010615
InChI Key GRPWQLDSGNZEQE-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
PubChem CID 75259
IUPAC Name 2-chloro-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Cl)C(=O)O

Specifications

Melting Point 183°C to 187°C
Quantity 25 g
Beilstein 1946215
Formula Weight 174.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.