missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzylamine, 97%

Product Code. 11387816
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387816 1 g 1g
11397816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387816 Supplier Thermo Scientific Alfa Aesar Supplier No. L12957.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 15205-11-5 | C7H7ClFN | 159.588 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15205-11-5
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD00042532
InChI Key CBKWAXKMZUULLO-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide
PubChem CID 139909
IUPAC Name (2-chloro-4-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)Cl)CN

Specifications

Boiling Point 94°C to 95°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.535
Quantity 1 g
UN Number UN2735
Beilstein 2936421
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.