Learn More
2-Chloro-4-fluorobenzylamine, 97%
Product Code. 11397816
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397816 5 g 5g
11387816 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397816 Supplier Thermo Scientific Alfa Aesar Supplier No. L12957.06
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 15205-11-5 | C7H7ClFN | 159.588 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15205-11-5
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD00042532
InChI Key CBKWAXKMZUULLO-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide
PubChem CID 139909
IUPAC Name (2-chloro-4-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)Cl)CN
Specifications
Boiling Point 94°C to 95°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.535
Quantity 5 g
UN Number UN2735
Beilstein 2936421
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzylamine