missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-fluoropyridine, 97%

Product Code. 11384416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384416 1 g 1g
11394416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384416 Supplier Alfa Aesar™ Supplier No. L20023.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 34941-91-8
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04039345
InChI Key FGSAQRJRWCZLOB-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
PubChem CID 2782792
IUPAC Name 2-chloro-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Cl

Specifications

Boiling Point 140°C
Flash Point 54°C (129°F)
Quantity 1 g
UN Number UN2929
Beilstein 1634747
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.