Learn More
Alfa Aesar™ 2-Chloro-4-fluoropyridine, 97%
Product Code. 11394416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394416 5 g 5g
11384416 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11394416 Supplier Alfa Aesar™ Supplier No. L20023.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 34941-91-8
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04039345
InChI Key FGSAQRJRWCZLOB-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
PubChem CID 2782792
IUPAC Name 2-chloro-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Cl
Specifications
Boiling Point 140°C
Flash Point 54°C (129°F)
Quantity 5 g
UN Number UN2929
Beilstein 1634747
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoropyridine