missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-iodo-3-pyridinemethanol, 97%, Thermo Scientific Chemicals

Product Code. p-7033044
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474440 250 mg 250mg
11484440 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474440 Supplier Thermo Scientific Alfa Aesar Supplier No. H50052.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 884494-44-4
Molecular Formula C6H5ClINO
Molecular Weight (g/mol) 269.466
MDL Number MFCD03095296
InChI Key BZQRQPAKTCMYIA-UHFFFAOYSA-N
Synonym 2-chloro-4-iodopyridin-3-yl methanol, 2-chloro-4-iodo-3-pyridinemethanol, 2-chloro-3-hydroxymethyl-4-iodopyridine, 2-chloro-4-iodo-pyridin-3-yl-methanol, acmc-20anzo, pubchem17089, 3-pyridinemethanol,2-chloro-4-iodo, 2-chloro-4-iodo-3-pyridyl methan-1-ol
PubChem CID 40418628
IUPAC Name (2-chloro-4-iodopyridin-3-yl)methanol
SMILES C1=CN=C(C(=C1I)CO)Cl

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 269.47
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-iodo-3-pyridinemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.