missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylbenzeneboronic acid, 95%

Product Code. 11472090
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472090 5 g 5g
11462090 1 g 1g
11482090 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472090 Supplier Thermo Scientific Alfa Aesar Supplier No. H28604.06
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 145349-62-8 | C7H8BClO2 | 170.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145349-62-8
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.399
MDL Number MFCD03411936
InChI Key UKYCKUPXBPLXBA-UHFFFAOYSA-N
Synonym 2-chloro-4-methylphenyl boronic acid, 2-chloro-4-methylbenzeneboronic acid, boronic acid, 2-chloro-4-methylphenyl, pubchem1778, 2-chloro-4-methyl-phenyl boronic acid, acmc-209cuq, ksc524i9h, 2-chloro-p-tolylboronic acid, 2-chloro-4-methyl phenyl boronic acid
PubChem CID 2773334
IUPAC Name (2-chloro-4-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C)Cl)(O)O

Specifications

Melting Point 198°C to 202°C
Quantity 5 g
Formula Weight 170.4
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-methylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.