Learn More
2-Chloro-4-nitrobenzamide, 98%
Product Code. 11486380
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486380 25 g 25g
11476380 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11486380 Supplier Thermo Scientific Alfa Aesar Supplier No. A19702.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 3011-89-0 | C7H5ClN2O3 | 200.578 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 3011-89-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD00017119
InChI Key GFGSZUNNBQXGMK-UHFFFAOYSA-N
Synonym aklomide, aklomix, clomide, alkomide, benzamide, 2-chloro-4-nitro, component of aklomix, component of novastat-w, unii-b0e341ra20, 2-chloro-4-nitro benzamide, dsstox_cid_21405
PubChem CID 2075
IUPAC Name 2-chloro-4-nitrobenzamide
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N
Spécification
Melting Point 168°C to 172°C
Quantity 25 g
Beilstein 1966537
Formula Weight 200.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrobenzamide