missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Product Code. 10712905 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10712905 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10712905 Supplier Thermo Scientific Acros Supplier No. 350210050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 69695-61-0
CAS Min % 94.0
Color Brown-Yellow to Yellow
CAS Max % 100.0
Density 98g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4790 to 1.4830
Assay Percent Range 94% min. (GC)
Molecular Formula C7H5ClF3NO
MDL Number MFCD01631704
Synonym 2-chloro-4-trifluoromethoxy aniline, benzenamine, 2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy phenylamine, benzenamine,2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy-phenylamine, pubchem2941, acmc-209wey, ksc630s5l, 2-chloro4-trifluoromethoxyaniline, 4-trifluoromethoxy-2-chloroaniline
InChI Key YMLWONHDNGICOH-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1OC(F)(F)F)Cl)N
IUPAC Name 2-chloro-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol) 211.57
PubChem CID 2734084
Formula Weight 211.57
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-trifluoromethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.