missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)pyrimidine, 97%

Product Code. 15498078
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498078 1 g 1g
15408088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498078 Supplier Alfa Aesar™ Supplier No. H61522.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 33034-67-2
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
MDL Number MFCD00115111
InChI Key FZRBTBCCMVNZBD-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl pyrimidine, pyrimidine, 2-chloro-4-trifluoromethyl, 4-trifluoromethyl-2-chloropyrimidine, 2-chloro-4-trifluoromethyl-pyrimidine, pubchem6907, acmc-209hxy, ksc222e4j, abbypharma ap-10-1130
PubChem CID 2773912
IUPAC Name 2-chloro-4-(trifluoromethyl)pyrimidine
SMILES C1=CN=C(N=C1C(F)(F)F)Cl

Specifications

Boiling Point 60°C (10 mmHg)
Flash Point 49°C (120°F)
Quantity 1 g
UN Number UN1993
Formula Weight 182.53
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.