missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyridine, 98%

Product Code. 11440838
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440838 25 g 25g
11420838 1 g 1g
11430838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440838 Supplier Thermo Scientific Alfa Aesar Supplier No. B25271.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69045-79-0 | C5H3ClIN | 239.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69045-79-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD01863635
InChI Key QWLGCWXSNYKKDO-UHFFFAOYSA-N
Synonym 2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
PubChem CID 4397176
IUPAC Name 2-chloro-5-iodopyridine
SMILES C1=CC(=NC=C1I)Cl

Specifications

Melting Point 95°C to 98°C
Quantity 25 g
Beilstein 108889
Sensitivity Light sensitive
Formula Weight 239.44
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-iodopyridine
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.