missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzaldoxime, 97%

Product Code. 11323656
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323656 5 g 5g
11313656 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323656 Supplier Thermo Scientific Alfa Aesar Supplier No. L14719.06
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 443-33-4 | C7H5ClFNO | 173.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-33-4
Molecular Formula C7H5ClFNO
Molecular Weight (g/mol) 173.57
MDL Number MFCD00837704
InChI Key OBJHLLOVMKKXDI-ONNFQVAWSA-N
Synonym 2-chloro-6-fluorobenzaldehyde oxime, 2-chloro-6-fluorobenzaldoxime, 6-chloro-2-fluorophenyl hydroxyimino methane, pubchem2883, e-n-2-chloro-6-fluorophenyl methylidene hydroxylamine, 1e-2-chloro-6-fluorobenzaldehyde oxime, e-1-2-chloro-6-fluorophenyl-n-hydroxymethanimine, ne-n-2-chloro-6-fluorophenyl methylidene hydroxylamine, ne-n-2-chloranyl-6-fluoranyl-phenyl methylidene hydroxylamine
PubChem CID 7329318
SMILES O\N=C\C1=C(F)C=CC=C1Cl

Specifications

Melting Point 125°C to 127°C
Quantity 5 g
Beilstein 3247886
IUPAC Name (NE)-N-[(2-chloro-6-fluorophenyl)methylidene]hydroxylamine
Formula Weight 173.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzaldoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.