Learn More
2-Chloro-D-phenylalanine, 98%
Product Code. 15463667
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463667 250 mg 250mg
11307129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15463667 Supplier Thermo Scientific Alfa Aesar Supplier No. H27359.MD
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 80126-50-7 | C9H10ClNO2 | 199.63 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 80126-50-7
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00077920
InChI Key CVZZNRXMDCOHBG-UHFFFAOYNA-N
Synonym 2-chloro-d-phenylalanine, h-d-phe 2-cl-oh, d-2-chlorophenylalanine, 2r-2-amino-3-2-chlorophenyl propanoic acid, d-2-chlorophe, d-phenylalanine, 2-chloro, r-2-amino-3-2-chlorophenyl propanoic acid, pubchem11960, pubchem17991, d-2-chloro phenylalanine
PubChem CID 6951092
IUPAC Name (2R)-2-amino-3-(2-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=CC=C1Cl)C(O)=O
Specifications
Melting Point 211°C to 213°C
Quantity 250 mg
Formula Weight 199.64
Percent Purity 98%
Chemical Name or Material 2-Chloro-D-phenylalanine