missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-[4-chloro-2-(2-chlorobenzoyl)phenyl]acetamide, TRC

Product Code. 30364152
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364152 1 g 1g
30354525 50 mg 50mg
30360057 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364152 Supplier Toronto Research Chemicals Supplier No. TRCC2930101G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14405-03-9
Molecular Formula C15H10Cl3NO2
Molecular Weight (g/mol) 342.6
Synonym 2,4'-Dichloro-2'-(o-chlorobenzoyl)acetanilide, 2-Chloro-N-[4-chloro-2-(2-chlorobenzoyl)phenyl]acetamide, Ro 97-1274
IUPAC Name 2-chloro-N-[4-chloro-2-(2-chlorobenzoyl)phenyl]acetamide
SMILES ClCC(=O)Nc1ccc(Cl)cc1C(=O)c2ccccc2Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H10Cl3NO2/c16-8-14(20)19-13-6-5-9(17)7-11(13)15(21)10-3-1-2-4-12(10)18/h1-7H,8H2,(H,19,20)
Formula Weight 340.9777
Chemical Name or Material 2-Chloro-N-[4-chloro-2-(2-chlorobenzoyl)phenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.