missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-diethylacetamide, 97%

Product Code. 11474664
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474664 5 g 5g
11484664 25 g 25g
11494664 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474664 Supplier Thermo Scientific Alfa Aesar Supplier No. A13691.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2315-36-8 | C6H12ClNO | 149.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-36-8
Molecular Formula C6H12ClNO
Molecular Weight (g/mol) 149.618
MDL Number MFCD00000928
InChI Key CQQUWTMMFMJEFE-UHFFFAOYSA-N
Synonym n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide
PubChem CID 16844
IUPAC Name 2-chloro-N,N-diethylacetamide
SMILES CCN(CC)C(=O)CCl

Specifications

Boiling Point 123°C to 124°C (15 mmHg)
Flash Point 113°C (235°F)
Quantity 5 g
UN Number UN2810
Beilstein 605866
Formula Weight 149.62
Percent Purity 97%
Chemical Name or Material 2-Chloro-N,N-diethylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.