missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-dimethylnicotinamide, 97%

Product Code. 15453698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453698 250 mg 250mg
15443698 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453698 Supplier Thermo Scientific Alfa Aesar Supplier No. H57162.MD
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 52943-21-2 | C8H9ClN2O | 184.623 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52943-21-2
Molecular Formula C8H9ClN2O
Molecular Weight (g/mol) 184.623
MDL Number MFCD08593346
InChI Key QNZRJGJNLOMEGJ-UHFFFAOYSA-N
Synonym 2-chloro-n,n-dimethylnicotinamide, 2-chloro-n,n-dimethyl-nicotinamide, 2-chloropyridine-3-carboxylic acid n,n-dimethylamide, 2-chloro-n,n-dimethyl-pyridine-3-carboxamide, 2-chloro-n,n'-dimethylnicotinamide, 3-pyridinecarboxamide,2-chloro-n,n-dimethyl, 2-chloro 3-pyridyl-n,n-dimethylcarboxamide, n,n-dimethyl-2-chloro-3-pyridinecarboxamide
PubChem CID 10965214
IUPAC Name 2-chloro-N,N-dimethylpyridine-3-carboxamide
SMILES CN(C)C(=O)C1=C(N=CC=C1)Cl

Specifications

Melting Point 63°C to 65°C
Quantity 250 mg
Formula Weight 184.63
Percent Purity 97%
Chemical Name or Material 2-Chloro-N,N-dimethylnicotinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.