missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacrylonitrile, TRC

Product Code. 30357977
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357977 50 g 50g
30355887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357977 Supplier Toronto Research Chemicals Supplier No. TRCC36383050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-37-6
Molecular Formula C3 H2 Cl N
Molecular Weight (g/mol) 87.508
Synonym 2-Propenenitrile, 2-chloro-, Acrylonitrile, 2-chloro- (6CI,7CI,8CI), 2-Chloro-2-propenenitrile, 1-Chloro-1-cyanoethene, 1-Chloro-1-cyanoethylene, 1-Cyano-1-chloroethene, 1-Cyanovinyl chloride, 2-Chloroacrylonitrile, 2-Chloropropenenitrile, NSC 511714, α-Chloroacrylonitrile
IUPAC Name 2-chloroprop-2-enenitrile
SMILES ClC(=C)C#N

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H2ClN/c1-3(4)2-5/h1H2
Formula Weight 86.9876
Chemical Name or Material 2-Chloroacrylonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.