missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzophenone, TRC

Product Code. 17978784
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978784 50 g 50g
17919354 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17978784 Supplier Toronto Research Chemicals Supplier No. TRCC36457550G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5162-03-8
Molecular Formula C13 H9 Cl O
Molecular Weight (g/mol) 216.66
Synonym (2-Chlorophenyl)phenylmethanone, Clotrimazole Imp. E (EP), 2-Chlorobenzophenone
IUPAC Name (2-chlorophenyl)-phenylmethanone
SMILES Clc1ccccc1C(=O)c2ccccc2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H9ClO/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9H
Formula Weight 216.0342
Chemical Name or Material 2-Chlorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.