missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl phenyl sulfide, 98%

Product Code. 11430867
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430867 5 g 5g
11440867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430867 Supplier Thermo Scientific Alfa Aesar Supplier No. B23265.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5535-49-9 | C8H9ClS | 172.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5535-49-9
Molecular Formula C8H9ClS
Molecular Weight (g/mol) 172.67
MDL Number MFCD00013691
InChI Key QDXIHHOPZFCEAP-UHFFFAOYSA-N
Synonym 2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6
PubChem CID 21702
IUPAC Name 2-chloroethylsulfanylbenzene
SMILES ClCCSC1=CC=CC=C1

Specifications

Density 1.178
Boiling Point 245°C
Flash Point 104°C (220°F)
Refractive Index 1.583
Quantity 5 g
UN Number UN2922
Beilstein 1907116
Formula Weight 172.68
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl phenyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.