missing translation for 'onlineSavingsMsg'
Learn More

2-chloroisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10476482
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 10476482

Brand: Thermo Scientific Maybridge SPB06649EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6313-54-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H4ClNO2
MDL Number MFCD00191402
Quantity 10 g
Synonym 2-chloroisonicotinic acid, 2-chloro-4-pyridinecarboxylic acid, 2-chloro-isonicotinic acid, 2-chloro isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro, 2-chloro-4-pyridinyl carboxylic acid, 2-chlorisonicotins, pubchem2598, 2-chloroisonicotinic acid,, 4-carboxy-2-chloropyridine
InChI Key QXCOHSRHFCHCHN-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=NC(Cl)=C1
IUPAC Name 2-chloropyridine-4-carboxylic acid
Molecular Weight (g/mol) 157.55
PubChem CID 236985
Formula Weight 157.56
Percent Purity 97%
Chemical Name or Material 2-Chloroisonicotinic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.