missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinoyl Chloride, TRC

Product Code. 30365840
Click to view available options
Quantity:
2 g
Unit Size:
2g
This item is not returnable. View return policy

Product Code. 30365840

Brand: Toronto Research Chemicals TRCC3737002G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloronicotinoyl Chloride
CAS 49609-84-9
Molecular Formula C6H3Cl2NO
Quantity 2 g
Synonym 2-Chloro-3-pyridinecarbonyl Chloride, 2-Chloronicotinic Chloride, 2-Chloronicotinoyl Chloride, 2-Chloronicotinyl Chloride, 2-Chloronictinoyl Chloride, 2-Chloropyridine-3-carboxylic Acid Chloride
InChI Formula InChI=1S/C6H3Cl2NO/c7-5-4(6(8)10)2-1-3-9-5/h1-3H
SMILES ClC(=O)c1cccnc1Cl
IUPAC Name 2-chloropyridine-3-carbonyl chloride
Molecular Weight (g/mol) 176
Formula Weight 174.9592
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.