missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloronicotinoyl Chloride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloronicotinoyl Chloride |
| CAS | 49609-84-9 |
| Molecular Formula | C6H3Cl2NO |
| Quantity | 1 g |
| Synonym | 2-Chloro-3-pyridinecarbonyl Chloride, 2-Chloronicotinic Chloride, 2-Chloronicotinoyl Chloride, 2-Chloronicotinyl Chloride, 2-Chloronictinoyl Chloride, 2-Chloropyridine-3-carboxylic Acid Chloride |
| InChI Formula | InChI=1S/C6H3Cl2NO/c7-5-4(6(8)10)2-1-3-9-5/h1-3H |
| SMILES | ClC(=O)c1cccnc1Cl |
| IUPAC Name | 2-chloropyridine-3-carbonyl chloride |
| Molecular Weight (g/mol) | 176 |
| Formula Weight | 174.9592 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction