missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl Cyclopentyl Ketone, TRC

Product Code. 30363233
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363233 50 g 50g
30358237 10 g 10g
30365407 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363233 Supplier Toronto Research Chemicals Supplier No. TRCC37782050G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6740-85-8
Molecular Formula C12H13ClO
Molecular Weight (g/mol) 208.68
Synonym (2-Chlorobenzoyl)cyclopentane, Cyclopentyl 2-Chlorophenyl Ketone, Cyclopentyl o-Chlorophenyl Ketone, (2-Chlorophenyl)cyclopentylmethanone
IUPAC Name (2-chlorophenyl)-cyclopentylmethanone
SMILES Clc1ccccc1C(=O)C2CCCC2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C12H13ClO/c13-11-8-4-3-7-10(11)12(14)9-5-1-2-6-9/h3-4,7-9H,1-2,5-6H2
Formula Weight 208.0655
Chemical Name or Material 2-Chlorophenyl Cyclopentyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.