missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyrimidine-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10492254
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10492254 1 g Amber Glass Bottle 1g
10285604 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492254 Supplier Thermo Scientific Maybridge Supplier No. MO08498DA
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 149849-92-3
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD06739041
InChI Key YMDSUQSBJRDYLI-UHFFFAOYSA-N
Synonym 2-chloropyrimidine-4-carboxylicacid, 4-pyrimidinecarboxylic acid, 2-chloro, 4-carboxy-2-chloropyrimidine, 2-chloro-pyrimidine-4-carboxylic acid, 2-chloro-4-pyrimidinecarboxylic acid, pubchem21879, acmc-209d3c, ksc497g7f, 2-chloropyrimidin-4-carboxylic acid, 2-chloro-pyrimidine-4-carboxylicacid
PubChem CID 22135751
IUPAC Name 2-chloropyrimidine-4-carboxylic acid
SMILES C1=CN=C(N=C1C(=O)O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.54
Percent Purity 97%
Chemical Name or Material 2-Chloropyrimidine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.