missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroquinoline, 99%, Thermo Scientific™

Product Code. 11475417
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475417 Supplier Thermo Scientific Alfa Aesar Supplier No. B23443.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloroquinoline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It possess antimicrobial activity against a panel of bacterial and fungal strains. It reacts with benzotriazole, followed by cyclization with loss of nitrogen, gives the indolo[2,3-b]quinoline ring system for use in novel cytotoxic DNA topoisomerase II inhibitors.

Solubility
Soluble in ethanol, ether, methanol, and chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-62-4
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00006741
InChI Key OFUFXTHGZWIDDB-UHFFFAOYSA-N
Synonym 2-chloro-quinoline, quinoline, 2-chloro, chloroquinoline, unii-ux7rin8gew, quinoline, chloro, ccris 3977, ux7rin8gew, chioroquinoline, 2-chloro quinoline, pubchem7580
PubChem CID 11928
IUPAC Name 2-chloroquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)Cl

Specifications

Melting Point 34°C to 37°C
Density 1.23
Boiling Point 265°C to 267°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 5 g
Beilstein 112561
Solubility Information Soluble in ethanol,ether,methanol,and chloroform. Insoluble in water.
Formula Weight 163.61
Percent Purity 99%
Chemical Name or Material 2-Chloroquinoline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.