missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotrityl Chloride (90%), TRC

Product Code. 30358605
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358605 1 g 1g
30354017 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358605 Supplier Toronto Research Chemicals Supplier No. TRCC4222601G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42074-68-0
Molecular Formula C19 H14 Cl2
Molecular Weight (g/mol) 313.22
Synonym 1-Chloro-2-(chlorodiphenylmethyl)benzene, Clotrimazole Imp. C (EP), Benzene, 1-chloro-2-(chlorodiphenylmethyl)-, 2-Chlorophenyldiphenylchloromethane, 2-Chlorophenyldiphenylmethyl chloride, Chloro(2-chlorophenyl)diphenylmethane, Diphenyl-2-chlorophenylmethyl chloride, o-Chlorotriphenylchloromethane
IUPAC Name 1-chloro-2-[chloro(diphenyl)methyl]benzene
SMILES Clc1ccccc1C(Cl)(c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H14Cl2/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H
Formula Weight 312.0473
Chemical Name or Material 1-Chloro-2-(chlorodiphenylmethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.