missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-ol, TRC

Product Code. 30366456
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366456 50 mg 50mg
30358695 100 mg 100mg
30364520 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366456 Supplier Toronto Research Chemicals Supplier No. TRCC99781550MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-67-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
IUPAC Name cyclohex-2-en-1-ol
SMILES OC1CCCC=C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h2,4,6-7H,1,3,5H2
Formula Weight 98.0732
Chemical Name or Material 2-Cyclohexen-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.