Learn More
2-Cyclohexylidene-cyclohexanone, TRC
Product Code. 30368539
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368539 250 mg -
30367244 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30368539 Supplier Toronto Research Chemicals Supplier No. TRCC995700250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1011-12-7
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.27
Synonym 2-Cyclohexylidenecyclohexanone, Dianon, Bicyclohexyliden-2-one
IUPAC Name 2-cyclohexylidenecyclohexan-1-one
SMILES O=C1CCCCC1=C2CCCCC2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C12H18O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H2
Formula Weight 178.1358
Chemical Name or Material 2-Cyclohexylidene-cyclohexanone